Learn More
Fisherbrand™ Low-Retention Pipette Tips, Nonfilter
Increase accuracy by reducing sample retention
 View more versions of this product
Catalog No. 02-717-141
Add to cart
Description
Description
  • Low retention surface minimizes samples loss
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Natural color
Specifications
Specifications
Universal Fit Pipettor
Beveled
Sterile
Low Retention
Hinged Rack
200μL
Natural
Universal Fit Pipettor
Documents
Accessories