Learn More
Fisherbrand™ Low-Retention Pipette Tips, Nonfilter Available on GSA/VA Contract for Federal Government customers only.
Increase accuracy by reducing sample retention
 View more versions of this product
Catalog No. 02-717-142
  Check Availability
Add to cart
Description
Description
  • Low retention surface minimizes samples loss
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Natural color
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Universal Fit Pipettor
Universal Fit
Non-sterile
Low retention
Bulk
200μL
Natural
Universal Fit Pipettor
Documents
Accessories