Fisherbrand™ Low-Retention Pipette Tips, Nonfilter

Increase accuracy by reducing sample retention

 View more versions of this product

Catalog No. 02-717-152

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Volume (Metric) 1000μL
    Sterility Nonsterile
    Color Natural
    Model Low Retention
    Tip Style Extended Length
    Packaging Stack Rack
    For Use With (Equipment) Universal Fit

    • Low retention surface minimizes samples loss
    • Certified by independent laboratory
    • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
    • Natural color

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit