Learn More
Fisherbrand™ Magnetic Stir Bars with Vanes Available on GSA/VA Contract for Federal Government customers only.
Ensure thorough sample mixing in test tubes, micro vials and conical-bottom centrifuge tubes.
 View more versions of this product
Catalog No. 14-513-71
Add to cart
Description
Description
Ensure thorough sample mixing in test tubes, micro vials and conical-bottom centrifuge tubes.
  • Made of solid FDA grade, USP Class VI Teflon™ and has a cross-mounted, molded-in Alnico magnet
  • Manufactured to fit 9.7/6.1 x 23.0 x 9.9mm tubes; can be trimmed to fit if needed without affecting the magnet
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yes
PTFE
10.4 x 16.5 x 9.8mm
50
10.4mm
Triangular
ISO 9001:2008
White
3 to 5mL vials and small conical-shaped tubes
Stir bar
Alnico Magnet