Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Marking Pens
Shop All Fisherbrand Products

Click to view available options
Color:
Black
Blue
Red
Description
- Fast drying—highly resistant to abrasion and water
- Designed to write on cold and damp surfaces as easily as they write on dry surfaces
- Will not smear when subjected to water or alcohol during normal use
- Ink is permanent on plastic, wood, and ceramic
- Washes off glass, metal, or porcelain surfaces
Order Info
Sold in packs of 10
Specifications
Specifications
| Product Type | Lab Permanent Marker |
| Tip Style | Fine |
| Color | Black |
| Sterility | Non-sterile |
| Latex Free | Yes |
| For Use With (Application) | Used to make permanent markings on plastic, metal, or porcelain labware |
| Quantity | 10 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction