Learn More
Fisherbrand™ Maxi Pipet Tips
For use with large volume pipets
 View more versions of this product
Catalog No. 02-707-132
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
For use with large volume pipets
  • Designed for use when transferring large sample volume
  • Advanced molding technology ensure tips are straight
  • Alternative option to serological pipets
  • Nonsterile; certified by independent laboratory to be RNase, DNase, DNA, and Pyrogen free
Specifications
Specifications
Rainin™ EDP2 Plus, Rainin™ EDP Plus
Universal Fit, Macro
Non-sterile
Yes
4.2 in.
Rainin™ EDP2 Plus, Rainin™ EDP Plus
Rack
250 to 2500μL
Natural
PP (Polypropylene)
10.6cm