Learn More
Fisherbrand™ Media-Miser™ Disposable Petri Dishes
Space saving, stackable, and sterilized
Specifications
Disposable Disposable
Material Polystyrene
Packaging Cut-apart Polyethylene Sleeves
Quantity 20/Sleeve
Sterility Sterile
View More Specs
Products 1
Catalog Number Diameter (Metric) Height (Metric) Price Quantity & Availability  
Catalog Number Diameter (Metric) Height (Metric) Price Quantity & Availability  
08-757-11YZ
View Documents
35 mm 10 mm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
These polystyrene dishes are smaller than standard Petri dishes, so less media is used.
  • Clear tops for distortion-free viewing
  • Venting ribs prevent condensation
  • Lids fit snugly when turned over, to reduce the risk of aerial contamination
  • Radiation sterilized
35 x 10mm Dishes
  • Raised, straight ridge around the top permits high stacking with little or no slipping, sliding, or scratching
  • 60 x 15mm Dishes
    • Raised, straight ridge around the top permits high stacking with little or no slipping, sliding, or scratching
  • 95 x 15mm Dishes
    • Raised, beveled ridge around the top for easy removal from a stack
    Order Info
    Packaged in cut-apart polyethylene sleeves, 20 per sleeve
    Specifications
    Specifications
    Disposable
    Cut-apart Polyethylene Sleeves
    Sterile
    Polystyrene
    20/Sleeve
    Yes