Learn More
Fisherbrand™ MicroDuster III Valve Assembly
The MicroDuster III Valve Assembly is a reusable, surgical grade assembly compatible with item number 2511. Features a chrome-plated brass valve with 3 ft. flexible extension hose.
Catalog No. 18-428
Add to cart
Description
Description
  • Removes duct particles from electronic equipment, keyboards, optical surface, precision instruments
  • No bitterant added
  • Nonflammable, 100% ozone safe
  • Contains HFC's
  • With 91.44cm (3 ft.) extension hose
Specifications
Specifications
Valve assembly
Valve and hose assembly for use with item number 2511