Learn More
Fisherbrand™ Motorized Pipet Fillers/Dispensers
Comfortable pipetting with precise control
 View more versions of this product
Catalog No. 03-692-168
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Yes
Two spare filters, autoclavable pipet adapter bench stand, AC recharger, space saving wall stand and instructions
120V
173g
Powerpette Plus
Glass or plastic pipet from 1 to 100mL
0.45μm
6 oz.
Pink
Pipet Filler
Accessories