Learn More
Fisherbrand™ Natural HDPE Wide Mouth Round Bottles with White Polypropylene PE Foam Caps
Click to view available options
Capacity (Metric):
3000 ml
Suggested for use with water based chemicals and some food products. Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles with White Polypropylene PE Foam Lined Cap feature a wide opening making them ideal for solids.
  • Ideal for general lab use with solids and powders.
  • Provide excellent moisture barrier
  • Impact resistant.
  • Ideal for high-speed filling, demanding pharmaceutical specifications and rough handling during shipping.
  • Translucent bottles allow limited viewing of contents
  • Provides moisture barrier and are impact resistantHigh Density Polyethylene with good chemical resistance
  • Round wide mouth style ideal for solids
Closure Size 100-400 mm
Mouth Wide
Closure Material Polypropylene
Closure Included Yes
Closure Liner Polyethylene Foam
Capacity (Metric) 3000 ml
Color Natural
Material HDPE
Quantity 48
Type Wide Mouth Round Bottle
Closure Color White
Neck Finish 100-400
Show More Show Less