Learn More
Fisherbrand™ Oil-Only Pads and Booms
Comprised of multilayer, fine fiber polypropylene for the most cost-effective sorption
Description
- Selectively remove hydrocarbons from water
- Utilize up to eight layers of fine fibers for superior wicking
- Spilled liquid adheres to the surface of and in between the numerous, multilayered fine fibers
- Use for smaller spills on land or water, for leaks from machinery and containers, and for wiping
- Bonded every 4 in. (10.2cm)
- Perforated vertically to fit in tight, narrow spaces
- Surround and contain spills on water
- Designed with strategically placed hooks to create boom-chains of any length
- Continue to float when fully saturated
- Contained in white fish netting sleeve for greater oil penetration and easy indication when saturation has been reached
Specifications
White | |
68.1 L | |
Oil Sorbent Boom | |
10 ft. x 5 in. (3.04 m x 12.7 cm) | |
3.04 m | |
12.7 cm |
18 gal. | |
Polypropylene | |
Designed with strategically placed hooks to create boom-chains of any length | |
10 ft. | |
5 in. |