Fisherbrand™ Oil-Selective Sorbents

Selectively remove hydrocarbons from water

 View more versions of this product

Catalog No. 17-215D

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type Oil Sorbent Sock
    Material Polypropylene
    Capacity (English) 6 gal.
    Capacity (Metric) 22.7L
    Length (Metric) 40.6cm
    Length (English) 16 in.
    For Use With (Application) For sorbing oil and a wide variety of acids and other hazardous chemicals
    Weight Capacity Double
    Width (English) 16 in.
    Width (Metric) 40.6cm
    Capacity 6 gal., 22.7L
    Dimensions (L x W) 16 x 16 in. (40.6 x 40.6cm)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit