Learn More
Fisherbrand™ Pasteur Pipets Available on GSA/VA Contract for Federal Government customers only.
Being packaged in strong corrugated material provides maximum protection from external shock for Fisherbrand™ Pasteur Pipets. Each pack also contains a foam pad to protect the pipet tips.
 View more versions of this product
Catalog No. 22-209361
Add to cart
Description
Description
Being packaged in strong corrugated material provides maximum protection from external shock for Fisherbrand™ Pasteur Pipets. Each pack also contains a foam pad to protect the pipet tips.
  • Straight-cut tips and smooth, fire-polished tops to facilitate bulb attachment
  • Positive constriction approximately 1 in. from the top to prevent cotton plugs from slipping into pipet barrel
  • Easel configuration of each pack keeps pipets orderly and easily accessible
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0.27 in.
9 in.
Straight
Easel
Soda Lime Glass
7 mm
Case of 4 x 250
Smooth fire polished
22.86 cm
2 mL
Documents
Promotions
Promotions
  • Fisherbrand , Fisher Scientific , DWK Life Sciences (Kimble)Supplier Diversity Partner
    Get up to $500 in Fisherbrand glassware at no additional cost with your qualifying glassware purchase.
    06-30-2021