Learn More
Fisherbrand™ Black Phenolic PolyCone Cap Available on GSA/VA Contract for Federal Government customers only.
Forms an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.
 View more versions of this product
Catalog No. 02-912-055
  Check Availability
Add to cart
Description
Description
PolyCone Lined Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier. This liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids, to soaps, to macro-invertebrates.
  • Liner molds itself around the sealing areas of finish as cap is screwed on
  • Eliminates leakage, evaporation, contamination, binding and backing off
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Phenolic
Black
576/Cs.
Black
PolyCone
Screw
Polyseal
PhenolicþPhenolic
Cap, PE Cone LinedþPolyCone Lined Cap
24/400
24-400