Fisherbrand™ Periodic Charts Products for Science Education

 View more versions of this product

Catalog No. 05-702-10

  Check Availability
Add to Cart

    Specifications

    Product Type Periodic chart
    Classification Chemistry
    Format Notebook size
    Height (English) 10.75 in.
    Includes Symbols, atomic numbers, atomic weights, element groups and maximum orbital electron distribution for each period
    Width (English) 8.25 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit