Learn More
Fisherbrand™ PFA Safety Coated Liquid-In-Glass Partial Immersion Thermometers
Added safety and protection from breakage
 View more versions of this product
Catalog No. 13-201-605
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
±1.5°C
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Partial
200mm
2°F
Organic
ISO/IEC 17025:2005, A2LA, NIST
50mm
7.9 in.
White Back Glass
20° to 230°F