Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ pH Paper Rolls Products for Science Education

General purpose pH papers in efficient 15' rolls

$29.00 - $35.25

Specifications

For Use With (Application) Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications
Storage Requirements Room Temperature
Quantity 100 Tests
View More Specs

Products 6
Catalog Number Type pH Range Price Quantity & Availability  
Catalog Number Type pH Range Price Quantity & Availability  
S22500 Products for Science Education
View Documents
pH Test Paper 0 to 3
Case of 5 for $29.00
Add to cart
 
S22501 Products for Science Education
View Documents
pH Test Paper 3 to 5.5
Case of 5 for $29.00
Add to cart
 
S22502 Products for Science Education
View Documents
pH Test Paper 6 to 8
Case of 5 for $29.00
Add to cart
 
S22503 Products for Science Education
View Documents
pH Test Paper 8 to 9.5
Case of 5 for $29.00
Add to cart
 
S22506 Products for Science Education
View Documents
pH Test Paper 1 to 12
Pack of 5 for $29.00
Add to cart
 
S22507 Products for Science Education
View Documents
pH Test Paper Set 0 to 14
Pack of 7 for $35.25
Add to cart
 
Description

Description

  • Convenient dispenser allows you to select the right amount of pH paper for each application
  • Easy-to-usedash (em)Dip paper in solution to be tested and match to color chart immediately
  • Highly accurate and well suited for most applications
  • Each SKU is packed in box of five clear plastic dispensers
  • Cat. No. 13-640-507 is a pH paper testing set that includes shrink wrapped pack of seven dispensers (one of each SKU)
  • Store at room temperature. Keep out of direct sunlight.

Order Info

15 ft. Roll provides about 100 tests.

Specifications

Specifications

Convenient dispenser allows to select the right amount of pH paper for each application, highly accurate and well suited for most applications
Room Temperature
100 Tests
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit