Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Pigskin Driver's Gloves Get IT Item

Return to original pliability even after being soaked in water

 View more versions of this product

Catalog No. 19-043020



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

10 x 5 in. (25.5 x 12.75cm)
Brown
Leather, pigskin
5 in.
0.9mm
Natural
10 in.
Slip-on
use where all Leather gloves are desired, extended wear is expected and superior worker satisfaction is demanded
Large
12.75cm
Powdered
Sterile
25.5cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit