Fisherbrand™ Pipette-Specific Tips
 View more versions of this product
Catalog No. 02-707-120
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type PipetteTips
    Packaging Lift-off Rack, 10 x 96
    Length (English) 2.13 in.
    Length (Metric) 5.4cm
    For Use With (Equipment) Biohit Proline, Nichiryo, Oxford BenchMate, Pipetman, Corning Lambda, Mettler Toledo VoluMate,Thermo Scientific Finnpipette, Rainin Classic, Eppendorf Research
    Sterility Sterile
    Color Yellow
    Tip Style Specialty
    Volume (Metric) 5 to 300μL
  • Description & Specifications
Related Products
Product Content Feedback