Learn More
Fisherbrand™ Plain Glass Microscope Slides
Precleaned to be free of debris
 View more versions of this product
Catalog No. 02-678-1
  Check Availability
Add to cart
Description
Description
  • 1mm thick slides manufactured from highest purity, corrosion-resistant clear sheet glass fully annealed to minimize chipping
  • Slides have improved surface wettability and are guaranteed nonsticking
  • Edges are precision ground for smoothness
  • Packed in environmental transport pack and hermetically sealed in a protective film to keep out moisture