Fisherbrand™ Polyethylene Biohazard Autoclave Bags without Sterilization Indicator Available on GSA/VA Contract for Federal Government customers only.

Strong and leak-resistant

 View more versions of this product

Catalog No. 22-044563

  Check Availability
Add to Cart

    Specifications

    Width (English) 19 in.
    Includes Biohazard symbol and text in English and Spanish
    Dimensions (L x W) 23 x 19 in. (58 x 48cm)
    Length (English) 23 in.
    Autoclavable Yes
    Thickness 1.25mil
    Length (Metric) 58cm
    Width (Metric) 48cm
    Closure Type Twist tie
    Material Polyethylene
    Color Printed Red

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit