Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Polypropylene Beard Cover

Cool and comfortable

Specifications

Color White
Material Polypropylene
For Use With (Application) Excellent for use in food preparation and industries where body contaminants are a concern
Latex Free Yes
Seam Type Elastic
View More Specs

Products 2
Catalog Number Size Price Quantity & Availability  
Catalog Number Size Price Quantity & Availability  
19-098-918
Accessories View Documents
X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
19-098-917
Accessories View Documents
Regular This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Excellent for use in food preparation and industries where body contaminants are a concern.

Excellent for use in food preparation and industries where body contaminants are a concern.

  • Cool, nonflammable, nonparticulating Comfortable elastic seams
  • Latex-free
  • White

Specifications

Specifications

White
Excellent for use in food preparation and industries where body contaminants are a concern
Elastic
Polypropylene
Yes
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit