Fisherbrand™ Polypropylene Beard Cover

Cool and comfortable

 View more versions of this product

Catalog No. 19-098-918

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Color White
    Seam Type Elastic
    Material Polypropylene
    Size XL
    For Use With (Application) Excellent for use in food preparation and industries where body contaminants are a concern
    Latex Free Yes

    Excellent for use in food preparation and industries where body contaminants are a concern.

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit