Learn More
Fisherbrand™ Polypropylene Cylinders
Calibrated “To Contain/To Deliver”
 View more versions of this product
Catalog No. 08-570-21G
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • One-piece molded construction with permanently molded-on graduations
  • New resin formulation provides greater clarity
  • Exposure to temperatures above 85°C (185°F) may alter accuracy
  • Repeated autoclaving may alter accuracy
Specifications
Specifications
1000mL
±12.00mL
10mL
PP (Polypropylene)
10mL