Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Polypropylene Disposable Beakers
Click to view available options
Capacity (Metric):
30 mL
100 mL
150 mL
Description
- Highly resistant to strong acids, organic solvents and other common laboratory reagents
- Special “dripless” lip Rib construction permits stacking without binding Base allows beaker to sit flat during use
- Graduated in mL and oz.
Specifications
Capacity (Metric) | 150 mL |
Material | Polypropylene |
For Use With (Application) | Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents |
Includes | Spout |
Increments | 5 mL |
Type | Beaker |
Autoclavable | Autoclavable |
Disposable | Single-use |
Graduated | Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction