Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Polypropylene Disposable Beakers

Click to view available options
Capacity (Metric):
30 mL
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL

Unbreakable; can be thrown away after use or steam-autoclaved for reuse

  • Highly resistant to strong acids, organic solvents and other common laboratory reagents
  • Special “dripless” lip Rib construction permits stacking without binding Base allows beaker to sit flat during use
  • Graduated in mL and oz.

Autoclavable Autoclavable
Disposable Yes
Graduated Yes
Capacity (Metric) 250 mL
Material Polypropylene
For Use With (Application) Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents
Includes Spout
Increments 10 mL
Type Beaker