Learn More
Fisherbrand™ Polypropylene Graduated Specimen Containers
Ideal for specimen collection, transportation and storage
Description
- Autoclavable
- Graduated
- With or without nonautoclavable polyethylene snap lid
Order Info
Note: Sterile containers with lids are individually wrapped, and are sterile inside container. Nonsterile and sterile containers without lids are bulk packed.
Specifications
No | |
Polypropylene | |
8 oz. |
Yes | |
Non-sterile | |
237 mL |