Learn More
Fisherbrand™ Polypropylene Microtiter Tubes, Plain tubes; Nonsterile; Bagged Available on GSA/VA Contract for Federal Government customers only.
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates
 View more versions of this product
Catalog No. 02-681-377
  Check Availability
Add to cart
Description
Description
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates
Also suitable for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagents. Choose plain tubes or tubes with frosted writing area.
  • Superior-grade polypropylene tubes and packing racks are autoclavable and can be frozen to -80°C
  • Nonsterile tubes are certified RNase- and DNase-free
  • Sterile tubes are certified RNase-, DNase-, ATP-, DNA- and pyrogen-free
  • Frosted tubes feature writing area that wraps around tube collar
  • Capacity- 1.2mL
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Packed in bulk, or in convenient 8 x 12 compact tray packs or robotic racks.
Compact Tray Packs
  • Snug, smooth- fitting cover protects samples and enables stacking
  • With paper grid for easy indentification of contents
Robotic Racks
  • Specifically designed with 96-well footprint for robotic workstations
  • One-way lid design with alphanumeric grid enables easy sample identification
  • Condensation rings inside cover minimize cross-contamination
  • Tubes fit in straight, upright position
Specifications
Specifications
Non-sterile
Yes
Natural
HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagent
Frosted
RNase, DNase-free
1.2mL
Polypropylene
Bag