Learn More
Fisherbrand™ Polystyrene Beakers
Ideal for use as sample containers
 View more versions of this product
Catalog No. 087-32-120
Add to cart
Description
Description
Ideal for use as sample containers
  • 20mL size is useful in flame photometry
  • 5mL size can be used in atomic absorption spectroscopy and flame photometry
Specifications
Specifications
0.67 oz.
43mm
Beaker
PS (Polystyrene)
27mm
Yes
20mL