Fisherbrand™ Polyvinyl-Lined Polypropylene Caps

Catalog No. 02-911-853

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Closure Size 70/400
    Quantity 640/PK.
    Closure Material Plastic
    Closure Type screw cap
    Description Screw cap
    Liner Material Polyvinyl
    Material HDPE
    Certifications/Compliance ISO 9001
    Color White
    No. per Case 640/Cs.
    Closure Liner Poly/Vinyl
    For Use With Bottle
    Neck Finish 70-400

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit