Fisherbrand™ Polyvinyl-Lined Polypropylene Caps

Catalog No. 02-911-804

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Sterile Yes
    Closure Material Plastic
    Closure Size 20/400
    Closure Type screw cap
    Description Screw cap
    Liner Material Polyvinyl
    Quantity 576/PK.
    Material HDPE
    Certifications/Compliance ISO 9001
    Closure Liner Poly/Vinyl
    Color White
    For Use With Bottle
    Neck Finish 20-400
    No. per Case 576/Cs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit