Learn More
Fisherbrand™ Porcelain Hirsch-Type Funnels with Perforated Plates
Highly resistant to chemical attack
$42.20 - $51.10
Specifications
Material | Porcelain |
---|---|
Autoclavable | Autoclavable |
Includes | Fixed perforated plate |
Quantity | Each |
Wall Thickness | 2mm |
Description
Highly resistant to chemical attack
With fixed perforated plate.
- Autoclavable
- Glazed inside and outside with the exception of the rim
Specifications
Porcelain | |
Fixed perforated plate | |
2mm |
Autoclavable | |
Each | |
Hirsch Funnel |