Learn More
Fisherbrand™ Powder-Free Vinyl Examination Gloves Get IT Item
Durable with extra stretch and superior fit
 View more versions of this product
Catalog No. 19-041-190B
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Smooth
Ambidextrous
Vinyl
5 mil
Powder-free
Non-sterile
24.1cm
Beaded/Rolled
Yes
Small
ISO9001 QMS
Clear
9.5 in.