Fisherbrand™ Powder-Free Vinyl Examination Gloves Get IT Item
Durable with extra stretch and superior fit
 View more versions of this product
Catalog No. 19-041-190C
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Certifications ISO9001 QMS
    Exterior Finish Smooth
    Glove Cuff Style Beaded
    Hand Ambidextrous
    Length (English) 9.5 in.
    Latex Free Yes
    Length (Metric) 24.1cm
    Material Vinyl
    Powdered No
    Size Medium
    Sterile No
    Thickness 5mil
    Color Clear
  • Description & Specifications
Related Products
Product Content Feedback