Learn More
Fisherbrand™ Poxygrid™ Test Tube Racks Available on GSA/VA Contract for Federal Government customers only.
Chemical and corrosion-resistant steel wire coated with heat-cured epoxy for long life and easy cleaning
 View more versions of this product
Catalog No. 14-793-9
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Rack, Test Tube
Blue
9.6 x 4.3 x 3.25 in. (24.3 x 11 x 8.3cm)
8.3cm
9.6 in.
Epoxy Coated Steel
4.3 in.
Yes
20mm
3.25 in.
Tubes (16-20mm)
24.3cm
40
11cm