Learn More
Fisherbrand™ Poxygrid™ Test Tube Racks Available on GSA/VA Contract for Federal Government customers only.
Chemical and corrosion-resistant steel wire coated with heat-cured epoxy for long life and easy cleaning.
 View more versions of this product
Catalog No. 14-793-15
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Chemical and corrosion-resistant steel wire coated with heat-cured epoxy for long life and easy cleaning.
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
  • All racks have square holes, three decks, a slotted grid bottom and reinforced edges
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Autoclavable
13 mm
2.5 in.
10 to 13 mm Tubes
20.5 cm
108
16.4 cm
Blue
8.1 x 6.5 x 2.5 in. (20.5 x 16.4 x 6.4cm)
6.4 cm
8.1 in.
Epoxy Coated Steel
6.5 in.
Test Tube Rack