Learn More
Fisherbrand™ Poxygrid™ Test Tube Racks
Chemical and corrosion-resistant steel wire coated with heat-cured epoxy for long life and easy cleaning.
 View more versions of this product
Catalog No. 14-793-4
Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
Chemical and corrosion-resistant steel wire coated with heat-cured epoxy for long life and easy cleaning.
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
  • All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Specifications
Autoclavable
Tubes (25 to 30 mm)
48
10.88 x 8.13 x 4 in. (28 x 21 x 10cm)
10.88 in.
8.13 in.
4 in.
30 mm
Blue
Epoxy Coated Steel
Test Tube Rack
28 cm
21 cm
10 cm
30 mm