Learn More
Fisherbrand™ Poxygrid™ Test Tube Racks
Chemical and corrosion-resistant steel wire coated with heat-cured epoxy for long life and easy cleaning.
 View more versions of this product
Catalog No. 14-793-12
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
Chemical and corrosion-resistant steel wire coated with heat-cured epoxy for long life and easy cleaning.
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
  • All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Specifications
Test Tube Rack
Blue
Epoxy Coated Steel
9.5 x 5.1 x 2.5 in. (24.2 x 13 x 6.4cm)
9.5 in.
5.1 in.
2.75 in.
16 mm
Autoclavable
Tubes (13 to 16 mm)
72
24.2 cm
13 cm
7 cm
16 mm
Product Suggestions
Product Suggestions