Learn More
Fisherbrand™ Prealigned Deuterium Lamps
With Spectrosil™ quartz or UV glass windows and All have 28 to 30mm O.D.
$953.00
Alert:
For safe operation of deuterium lamps, the operator should never look directly at the lamp. Products produce high levels of deep UV light, and it is imperative that appropriate eye protection is used. Also, the envelope can reach 200°C during operation, and proper precautions must be taken if lamp is to be handled.
Products 1
Catalog Number For Use With (Equipment) Price Quantity & Availability  
Catalog Number For Use With (Equipment) Price Quantity & Availability  
14-386-114
View Documents
Alpkem 510/5010, Altex 153, Bio-Rad Biofocus 2000/3000, Carlo Erba UV/Vis 20, 20RS, Dionex UVM-1, DSA-1, Linear 200/203/204/205/206/205PH
Each for $953.00
Add to cart
Description
Description
  • With Spectrosil™ quartz or UV glass windows
  • All have 28 to 30mm O.D.