Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Premium Insulated Leather Drivers Gloves: Foam Lined

Made from top-grain cowhide leather

$103.00 - $107.00

Specifications

Color Natural
Glove Cuff Style Open
Grade Premium
Liner Material Foam
Material Top Grain Cowhide Leather
View More Specs

Products 4
Catalog Number Size Price Quantity & Availability  
Catalog Number Size Price Quantity & Availability  
19-013378
View Documents
Medium
Dozen of 12 for $103.00
Case of 6 DZ for $618.00
Add to cart
 
19-013379
View Documents
Large
Dozen of 12 for $103.00
Case of 6 DZ for $618.00
Add to cart
 
19-013381
View Documents
X-Large
Dozen of 12 for $107.00
Case of 6 DZ for $642.00
Add to cart
 
19-013374
View Documents
Small This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Ideal for use in cold weather applications including farming, construction, machine operation, and refrigeration.

  • Straight thumb and shirred elastic back
  • Red foam lining adds comfort and warmth
  • Gunn-cut style
  • Easy size identification by color and cuff

Specifications

Specifications

Natural
Premium
Top Grain Cowhide Leather
Gunn
Open
Foam
Drivers Gloves
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit