Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Premium Insulated Leather Driver's Gloves: Foam Lined

Made from top-grain cowhide

 View more versions of this product

Catalog No. 19-013378

  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

9.7 x 4.8 in. (24.75 x 12.25cm)
Green
Leather, cowhide
Medium
12.25cm
Powdered
Sterile
24.75cm
Slip-on
cold weather applications including farming, construction, machine operation and refrigeration
Foam
4.8 in.
0.7 to 0.9mm
Natural
9.7 in.
Promotions

Promotions

  • Keep yourself and your lab safe with these great deals.

    12-31-2017

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit