Learn More
Fisherbrand™ Premium Plus Robotic Tips
BioGuard Aerosol Barriers prevent contamination of robot head and cross-contamination of samples
 View more versions of this product
Catalog No. 05-408-165
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Color-coded for easy size identification
  • Reinforced racks provide sturdy platform for loading tips
  • Available in all robot-supported volumes
  • Reliable alternative to expensive OEM tips
  • Available with Low Binding Surface Tension technology
  • 100% QC for straight tips
Order Info
  • Biomek 1000/2000 tips fit MP200 head (250µL tips) and MP20 head (20µL tips)
  • Biomek FX/ Multimek™ AP96 tips fit AP96 head on Biomek FX and Multimek
  • Supplied in racks of 96, 100 racks per pack, five packs per case
Specifications
Specifications
Universal Fit
Sterile
Natural