Learn More
Fisherbrand™ Premium Plus Robotic Tips
BioGuard Aerosol Barriers prevent contamination of robot head and cross-contamination of samples
Products 13
Catalog Number Description Price Qty Availability  
Catalog Number Description Price Qty Availability  
05-408-168 View Documents N/A Vol: 1 - 20µL; Biomek FX/Multimek AP96; Standard; w/BioGuard; Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-160 View Documents N/A Vol: 1 - 20µL; Biomek 1000/2000; Standard; Blue; Nonsterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-163 View Documents N/A Vol: 1 - 250µL; Biomek 1000/2000; Standard; Natural; Nonsterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-164 View Documents N/A Vol: 1 - 250µL; Biomek 1000/2000; Standard; Natural; Sterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-162 View Documents N/A Vol: 1 - 20µL; Biomek 1000/2000; Standard; w/BioGuard; Blue; Nonsterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-167 View Documents N/A Vol: 1 - 20µL; Biomek FX/Multimek AP96; Standard; Blue; Sterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-165 View Documents N/A Vol: 1 - 170µL;Biomek 1000/2000; Standard; w/BioGuard; Natural; Sterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-170 View Documents N/A Vol: 1 - 250µL; Biomek FX/Multimek AP96; Standard; Natural; Sterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-172 View Documents N/A Vol: 1 - 20µL; Biomek 1000/2000; Low binding; Blue; Nonsterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-179 View Documents N/A Vol: 1 - 20µL; Biomek FX/Multimek AP96; Low binding; Blue; Sterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-180 View Documents N/A Vol: 1 - 20µL; Low Binding; w/BioGuard; Sterile rack, Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-181 View Documents N/A Vol: 1 - 250µL; Biomek FX/Multimek AP96; Low binding; Natural; Nonsterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-408-182 View Documents N/A Vol: 1 - 250µL; Biomek FX/Multimek AP96; Low binding; Natural; Sterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Color-coded for easy size identification
  • Reinforced racks provide sturdy platform for loading tips
  • Available in all robot-supported volumes
  • Reliable alternative to expensive OEM tips
  • Available with Low Binding Surface Tension technology
  • 100% QC for straight tips
Order Info
  • Biomek 1000/2000 tips fit MP200 head (250µL tips) and MP20 head (20µL tips)
  • Biomek FX/ Multimek™ AP96 tips fit AP96 head on Biomek FX and Multimek
  • Supplied in racks of 96, 100 racks per pack, five packs per case