Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand PTFE Stirring Rods
Pure PTFE rods are available either solid or with a milled steel core.
Description
Pure PTFE rods are available either solid or with a milled steel core.
- Solid rods have tapered ends
- Steel-core rods can be bent into a permanent shape
- Both types are inert, will not scratch, and can be used at temperatures up to 280°C (536°F)
Order Info
Solid PTFE rods measure approximately 0.3125 in. O.D. (8mm); steel-core rods, 0.25 in. O.D. (6.35mm).
Specifications
ISO 9001:2008 | |
0.31 in. | |
12 | |
8mm | |
PTFE (Polytetrafluoroethylene) |
White | |
8 in. | |
Stirring Rod | |
203.2mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title