Fisherbrand™ PVC-Coated String Knit Gloves

 View more versions of this product

Catalog No. 19-053388

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Recommend washing gloves in slightly warm water not exceeding 40°C, a mild commercial laundry soap or detergent should be used, wash in a 5 to 10 min. cycle time, rinse in cold water, tumble dry at temperature not exceeding 60°C
    Glove Cuff Style String knit
    For Use With (Application) warehousing and package handling, general assembly, light duty general work situations, construction and bottle handling
    Length (English) 10 in.
    Length (Metric) 25.4cm
    Material PVC
    Liner Material Cotton, Polyester
    Powdered Yes
    Size Men's
    Sterile Yes
    Color White, blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit