Learn More
Fisherbrand™ PVC-Coated String Knit Gloves with Honeycomb Finish
Polyester shell dries quickly, making these gloves ideal for use in wet applications
$13.75
Specifications
Cleaning Procedure Recommend washing gloves in tempered water not exceeding 40°C, a mild commercial laundry soap or detergent should be used, wash in a 5 to 10 min. cycle time, rinse in cold water, hang dry
Exterior Finish Seamless
For Use With (Application) Glass, plywood and lumber handling and fishing
Glove Cuff Style Continuous knit
Liner Material Polyester
View More Specs
Products 1
Catalog Number Size Price Quantity & Availability  
Catalog Number Size Price Quantity & Availability  
19-816-928
View Documents
Medium
Dozen of 12 for $13.75
Case of 12 DZ for $165.00
Add to cart
Description
Description
  • Coated with PVC “honeycomb” pattern on both sides for extra grip and cut resistance
  • Hand-to-hand reversible in women's and men's universal sizes
  • Orange with color-coded cuff
Specifications
Specifications
Recommend washing gloves in tempered water not exceeding 40°C, a mild commercial laundry soap or detergent should be used, wash in a 5 to 10 min. cycle time, rinse in cold water, hang dry
Glass, plywood and lumber handling and fishing
Polyester
PVC
Seamless
Continuous knit
Orange