Fisherbrand™ Qualitative Grade Plain Filter Paper Circles and Sheets - P2 Grade

Ideal for clarifying and removing particles of 1µm and larger, and in applications that do not require a low-ash or ashless paper

 View more versions of this product

Catalog No. 09-803-5D

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Particle Retention 1 to 5μm
    Format Circle
    For Use With (Application) Ideal for clarifying and removing particles of 1μm and larger, and in applications that do not require a low-ash or ashless paper
    Porosity Fine
    Diameter (Metric) 11cm
    Grade P2

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit