Learn More
Fisherbrand™ Suprasil™ Quartz Micro Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Fisherbrand™ Suprasil™ Quartz Micro Cell Cuvettes are supplied with a PTFE cover.
 View more versions of this product
Catalog No. 14-385-927B
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Sold in pairs
  • Very chemically resistant
  • Maintains shape, even at high temperatures up to approximately 1000°C
  • 80% transmission from 320 to 2500nm
Specifications
Specifications
Quartz