Learn More
Fisherbrand™ Quartz, Silica and Glass Standard Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Fisherbrand™ Quartz, Silica and Glass Standard Cell Cuvettes provide optically excellent transparency.
 View more versions of this product
Catalog No. 14-385-902C
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Quartz - high UV transmission and low thermal expansion
  • Glass - used in the visible range of the spectrum
  • Low values of refractive index and dispertion
  • Good resistance to chemicals
Specifications
Specifications
Glass, Quartz, Silica