Learn More
Fisherbrand™ Racer Series Top Fuel Eyewear
Exceptional protection and style at an affordable price
This item is not returnable.
View return policy
Description
Exceptional protection and style at an affordable price
- Features a single-lens design with a sporty, dual-lens look
- Nose bridge and bayonet-style side arms grip for a comfortable, confident fit
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Meets ANSI Z87.1-2003 standards. | |
Colorful Mirror | |
Polycarbonate |
Scratch-resistant | |
Black |